SY3390162087 Synthonix Corporation

3-(Boc-amino)-4-trifluoromethylpyridine

  • MDL number MFCD22573369
  • Cas Number : 1392804-92-0
  • Molecular Weight : 262.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7398.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7398.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H335
Availability details for
Quantity
  • Availability
  • Add to Cart