SY3390162091 Synthonix Corporation

2-Aza-tricyclo[3.3.1.13,7]decane-5-carboxylic acid methyl ester hydrochloride

  • MDL number MFCD19707044
  • Cas Number : 1389264-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7304.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7304.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart