SY3390162130 Synthonix Corporation

5-(3,3-Difluorocyclobutyl)-2H-pyrazol-3-ylamine

  • MDL number MFCD22415228
  • Cas Number : 1425931-95-8
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7382.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7382.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H335
Availability details for
Quantity
  • Availability
  • Add to Cart