SY3390162133 Synthonix Corporation

10,11-dihydro-5H-Dibenzo[a,d]cycloheptene-5-methanol

  • MDL number MFCD22573382
  • Cas Number : 2975-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7832.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/7832.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart