SY3390162140 Synthonix Corporation

Ethyl 7-(tert-boc-amino)-9-azabicyclo[3.3.1]nonane-3-carboxylate

  • MDL number MFCD22573386
  • Cas Number : 1363380-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7375.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7375.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart