SY3390162144 Synthonix Corporation

Ethyl 7-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate

  • MDL number MFCD19707047
  • Cas Number : 1363382-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/7948.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7948.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart