SY3390162151 Synthonix Corporation

2-Hydrazino-1-methylpyridinium tosylate

  • MDL number MFCD09832907
  • Cas Number : 1404375-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7302.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7302.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart