SY3390162182 Synthonix Corporation

Oxo-(1-triisopropylsilyl-1H-pyrrol-3-yl)acetic acid ethyl ester

  • MDL number MFCD22573394
  • Cas Number : 87630-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/2018.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/2018.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart