SY3390162197 Synthonix Corporation

4-Amino-3-bromothieno[3,2-c]pyridine

  • MDL number MFCD08275115
  • Cas Number : 799293-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/8025.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8025.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart