SY3390162198 Synthonix Corporation

2-Amino-4-chloro-5-nitropyridine

  • MDL number MFCD04973545
  • Cas Number : 24484-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7951.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7951.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart