SY3390162233 Synthonix Corporation

tert-Butyl N-(trans-3-hydroxycyclobutyl)-N-methylcarbamate

  • MDL number MFCD21362306
  • Cas Number : 1033718-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8137.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/8137.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart