SY3390162275 Synthonix Corporation

(R)-2-Methylpyrrolidine-2-carboxylic acid

  • Cas Number : 63399-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7933.html

Safety Information

MSDS https://synthonix.com/7933.html
Availability details for
Quantity
  • Availability
  • Add to Cart