SY3390162302 Synthonix Corporation

7-Bromo-6-chloro-4-quinazolinone

  • MDL number MFCD10000555
  • Cas Number : 17518-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/7870.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/7870.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart