SY3390162331 Synthonix Corporation

(2S)-2-Amino-2-(oxetan-3-yl)acetic acid

  • MDL number MFCD19212929
  • Cas Number : 394653-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8466.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8466.html
hstatements H315 , H317 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart