SY3390162361 Synthonix Corporation

3-(3-Bromophenyl)oxetane

  • MDL number MFCD17012864
  • Cas Number : 1044507-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/8384.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8384.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart