SY3390162368 Synthonix Corporation

7-Bromo-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD08544270
  • Cas Number : 114744-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8109.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8109.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart