SY3390162369 Synthonix Corporation

3-Bromo-5-(trifluoromethyl)-1H-indazole

  • MDL number MFCD09998163
  • Cas Number : 1086378-32-6
  • Molecular Weight : 265.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/7797.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/7797.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart