SY3390162395 Synthonix Corporation

N-(3-bromo-2-methyl-6-nitrophenyl)-2,2,2-trifluoroacetamide

  • MDL number MFCD23703161
  • Cas Number : 1325729-87-0
  • Molecular Weight : 327.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(3-bromo-2-methyl-6-nitrophenyl)-2,2,2-trifluoroacetamide
Inchi Code 1S/C9H6BrF3N2O3/c1-4-5(10)2-3-6(15(17)18)7(4)14-8(16)9(11,12)13/h2-3H,1H3,(H,14,16)
Inchi Key SAZSMEKVWRGZIH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/9187.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/9187.html
pictograms GHS05, GHS06, GHS07
hstatements H301, H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart