SY3390162401 Synthonix Corporation

4,4-Dimethyl-3-oxo-pentanal

  • MDL number MFCD00042806
  • Cas Number : 23459-13-4
  • Molecular Weight : 128.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8624.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/8624.html
pictograms GHS05, GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart