SY3390162403 Synthonix Corporation

Ethyl 2-amino-6-Boc-4,7-dihydro-5H-thieno[3,2-c]pyridine-3-carboxylate

  • MDL number MFCD06796315
  • Cas Number : 193537-14-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/8060.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/8060.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart