SY3390162419 Synthonix Corporation

3-(3-Fluorophenyl)oxetan-3-amine hydrochloride

  • MDL number MFCD19381727
  • Cas Number : 1332839-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8584.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8584.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart