SY3390162468 Synthonix Corporation

2-Oxaspiro[3.5]nonane-7-methanol

  • MDL number MFCD22628753
  • Cas Number : 1256546-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/8486.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/8486.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart