SY3390162472 Synthonix Corporation

3-Oxetanethiol

  • MDL number MFCD19443271
  • Cas Number : 880136-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/8471.html

Safety Information

MSDS https://synthonix.com/8471.html
hstatements H302 ,
Availability details for
Quantity
  • Availability
  • Add to Cart