SY3390162476 Synthonix Corporation

[4-(Oxetan-3-yloxy)phenyl]methanamine

  • MDL number MFCD20233039
  • Cas Number : 1349719-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/8504.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/8504.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart