SY3390162480 Synthonix Corporation

2-(Oxetan-3-yloxy)pyrimidin-5-ylamine

  • MDL number MFCD20233082
  • Cas Number : 1349709-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8512.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/8512.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart