SY3390162491 Synthonix Corporation

1-(3-Oxetanyl)-1H-pyrazole-5-boronic acid pinacol ester

  • MDL number MFCD22628755
  • Cas Number : 1876473-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8591.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/8591.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart