SY3390162509 Synthonix Corporation

1,3,5-tris[Hydroxymethyl]benzene

  • MDL number MFCD01108484
  • Cas Number : 4464-18-0
  • Molecular Weight : 168.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8626.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/8626.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart