SY3390162526 Synthonix Corporation

Methyl 5-(aminomethyl)tetrahydrofuran-2-carboxylate hydrochloride

  • MDL number MFCD23152697
  • Cas Number : 1304126-25-7
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8646.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/8646.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart