SY3390344074 Synthonix Corporation

Monobenzalpentaerythritol

  • MDL number MFCD00085385
  • Cas Number : 2425-41-4
  • Molecular Weight : 224.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/8631.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/8631.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart