SY3390512681 Synthonix Corporation

1-(Azetidin-3-yl)-3-methylazetidin-3-ol dihydrochloride

  • MDL number MFCD22581551
  • Cas Number : 1403766-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/8836.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/8836.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart