SY3390512683 Synthonix Corporation

4-(Azetidin-3-yl)-1-methylpiperazin-2-one dihydrochloride

  • Cas Number : 1403766-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/8839.html

Safety Information

MSDS https://synthonix.com/8839.html
Availability details for
Quantity
  • Availability
  • Add to Cart