SY3390512711 Synthonix Corporation

2-(2-Aminopropan-2-yl)-N-(4-fluorobenzyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidine-4-carboxamide

  • Cas Number : 518048-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/8957.html

Safety Information

MSDS https://synthonix.com/8957.html
Availability details for
Quantity
  • Availability
  • Add to Cart