SY3390512770 Synthonix Corporation

2-Chloro-4-cyano-6-fluorobenzoic acid

  • Cas Number : 1258298-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/8773.html

Safety Information

MSDS https://synthonix.com/8773.html
Availability details for
Quantity
  • Availability
  • Add to Cart