SY3390512776 Synthonix Corporation

2-Chloro-4-methylpyrimidine-5-carboxylic acid

  • Cas Number : 188781-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/8877.html

Safety Information

MSDS https://synthonix.com/8877.html
Availability details for
Quantity
  • Availability
  • Add to Cart