SY3390512876 Synthonix Corporation

2-Methyl-4-pyrimidinamine

  • MDL number MFCD00626295
  • Cas Number : 74-69-1
  • Molecular Weight : 109.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methyl-4-pyrimidinamine
Inchi Code 1S/C5H7N3/c1-4-7-3-2-5(6)8-4/h2-3H,1H3,(H2,6,7,8)
Inchi Key GKVDLTTVBNOGNJ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form White to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/9010.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/9010.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart