SY3390512928 Synthonix Corporation

6-(Trifluoromethyl)pyrimidin-4-amine

  • MDL number No MDL found
  • Cas Number : 672-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/8991.html

Safety Information

MSDS https://synthonix.com/8991.html
Availability details for
Quantity
  • Availability
  • Add to Cart