SY3394381404 Synthonix Corporation

6-(4-Methylpiperazin-1-yl)pyrimidin-4-amine

  • Cas Number : 96225-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/9091.html

Safety Information

MSDS https://synthonix.com/9091.html
Availability details for
Quantity
  • Availability
  • Add to Cart