SY3394381480 Synthonix Corporation

tert-butyl 4-(4-Bromophenyl)sulfonylpiperazine-1-carboxylate

  • MDL number MFCD09746327
  • Cas Number : 259808-63-4
  • Molecular Weight : 405.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/9208.html

Safety Information

signalword Warning
pcodes P202, P261, P280
MSDS https://synthonix.com/9208.html
pictograms GHS07
hstatements H301, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart