SY3397819304 Synthonix Corporation

7-Tributylstannylchromen-2-one

  • MDL number MFCD25562898
  • Cas Number : 2098953-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/10004.html

Safety Information

pcodes P273 , P280 ,
MSDS https://synthonix.com/10004.html
hstatements H301 , H312 , H315 , H319 ,
Availability details for
Quantity
  • Availability
  • Add to Cart