SY3397819307 Synthonix Corporation

4-Iodo-6-methylsulfanyl-pyrimidine

  • MDL number MFCD25562902
  • Cas Number : 89125-96-2
  • Molecular Weight : 252.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/10030.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/10030.html
pictograms GHS06, GHS07, GHS08, GHS09
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart