SY3397819310 Synthonix Corporation

(2-Oxochromen-7-yl)boronic acid

  • MDL number MFCD25562900
  • Cas Number : 1357078-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/10014.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/10014.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart