SY3397819312 Synthonix Corporation

2-Oxo-2H-chromen-7-yl trifluormethanesulfonate

  • MDL number MFCD18382236
  • Cas Number : 108530-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/10002.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/10002.html
hstatements H302 , H315 , H318 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart