SY3397819313 Synthonix Corporation

2-(Phenylthio)acetamide

  • MDL number MFCD00040684
  • Cas Number : 22446-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/10024.html

Safety Information

pcodes P280 ,
MSDS https://synthonix.com/10024.html
hstatements H301 , H302 , H319 ,
Availability details for
Quantity
  • Availability
  • Add to Cart