SY3397819315 Synthonix Corporation

4-(Tributylstannyl)-2-(trimethylsilyl)thiazole

  • MDL number MFCD01319064
  • Cas Number : 252562-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 85
Country of Origin China
Product Link https://synthonix.com/5764.html

Safety Information

pcodes P261 , P273 , P280 ,
MSDS https://synthonix.com/5764.html
hstatements H301 , H312 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart