SY3405195509 Synthonix Corporation

5-Bromo-3-methylimidazo[1,2-a]pyridine hydrobromide

  • MDL number MFCD22422284
  • Cas Number : 74420-43-2
  • Molecular Weight : 291.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3373.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3373.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart