SY3405195510 Synthonix Corporation

2-Chloro-3,3-diethoxyprop-1-ene

  • MDL number MFCD23102091
  • Cas Number : 7575-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/3696.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/3696.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart