SY3408585597 Synthonix Corporation

4-Carboxy-Pennsylvania Green methyl ester

  • MDL number MFCD26406706
  • Cas Number : 879288-18-3
  • Molecular Weight : 396.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/1300.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/1300.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart