SY3408585598 Synthonix Corporation

[1-(azetidin-3-yl)-4-piperidyl]methanol di trifluoro acetic acid salt

  • MDL number MFCD26406707
  • Cas Number : 1651840-82-2
  • Molecular Weight : 398.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/10071.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/10071.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart