SY3408585606 Synthonix Corporation

4-(tert-Butyldimethylsilyloxy)butanal

  • MDL number MFCD12025097
  • Cas Number : 87184-81-4
  • Molecular Weight : 202.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3803.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3803.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart