SY3408585609 Synthonix Corporation

Dibromoacetaldehyde diethyl acetal

  • MDL number MFCD08457885
  • Cas Number : 761-17-1
  • Molecular Weight : 275.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/3669.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/3669.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart